Andrographolide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (S,E)-4-Hydroxy-3-(2-((1R,4aS,5R,6R,8aS)-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylenedecahydronaphthalen-1-yl)ethylidene)dihydrofuran-2(3H)-one

A2459-1G A2459-5G
TCA2459-1GEA 32 USD
TCA2459-1G TCA2459-5G
Andrographolide ≥98.0% (by HPLC)
Andrographolide

CAS Number: 5508-58-7
MDL Number: MFCD07778082
Molecular Formula: C20H30O5
Molecular Weight: 350.46
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 231
Specific rotation [a]20/D: -126 deg (C=1.5, AcOH)
Lambda max.: 223 nm (MeOH)

Formula: C20H30O5
MW: 350.45 g/mol
Melting Pt: 231 °C
MDL Number: MFCD07778082
CAS Number: 5508-58-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa4ee4784e351c28b8de7315504dbc37a4416270&vid=fa4ee4784e351c28b8de7315504dbc37a4416270&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR