1-Amino-2-methylpropan-2-ol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Amino-2-methyl-2-propanol, 1,1-Dimethylethanolamine

A2442-1G A2442-5G
TCA2442-1GEA 72 USD
TCA2442-1G TCA2442-5G
1-Amino-2-methylpropan-2-ol ≥98.0% (by GC)
1-Amino-2-methylpropan-2-ol

CAS Number: 2854-16-2
MDL Number: MFCD01697283
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: 1,1-Dimethylethanolamine

Formula: C₄H₁₁NO
MW: 89.1362 g/mol
Boiling Pt: 151 °C
Density: 0.94 (20 °C)
MDL Number: MFCD01697283
CAS Number: 2854-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4047aebd822751e0a0b422b216aff0a6ee03e454&vid=4047aebd822751e0a0b422b216aff0a6ee03e454&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR