2-Amino-6-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A2435-1G
A2435-5G
TCA2435-1GEA
23
USD
TCA2435-1G
TCA2435-5G
2-Amino-6-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
2-Amino-6-(trifluoromethyl)pyridine
CAS Number: 34486-24-3
MDL Number: MFCD03093868
Molecular Formula: C6H5F3N2
Molecular Weight: 162.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 87
Formula:
C₆H₅F₃N₂ MW: 162.11 g/mol Melting Pt: 87 °C |
MDL Number:
MFCD03093868 CAS Number: 34486-24-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...