2-Amino-6-(trifluoromethoxy)benzothiazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole

A2423-25G A2423-5G
TCA2423-25GEA 199 USD
TCA2423-25G TCA2423-5G
2-Amino-6-(trifluoromethoxy)benzothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-6-(trifluoromethoxy)benzothiazole

CAS Number: 1744-22-5
MDL Number: MFCD00210213
Molecular Formula: C8H5F3N2OS
Molecular Weight: 234.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 118


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Neurotransmitter Agent Ingredients, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: Riluzole

Formula: C₈H₅F₃N₂OS
MW: 234.20 g/mol
Melting Pt: 118 °C
MDL Number: MFCD00210213
CAS Number: 1744-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c3b160029a6a908e192925dc311b9f2aac670f4&vid=4c3b160029a6a908e192925dc311b9f2aac670f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR