3-Amino-5-methylpyridine ≥97.0%
Supplier: TCI America
Synonyms:
5-Amino-3-picoline
A2396-1G
A2396-5G
TCA2396-1GEA
37
USD
TCA2396-1G
TCA2396-5G
3-Amino-5-methylpyridine ≥97.0%
3-Amino-5-methylpyridine
CAS Number: 3430-19-1
MDL Number: MFCD04112508
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 155
Melting point (°C): 65
Other Product Names / Synonyms: 5-Amino-3-picoline
Formula:
C6H8N2 MW: 108.1411 g/mol Boiling Pt: 155 °C Melting Pt: 65 °C |
MDL Number:
MFCD04112508 CAS Number: 3430-19-1 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...