2-(2-Aminoethyl)-1-methylpyrrolidine ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
2-(1-Methyl-2-pyrrolidinyl)ethylamine
A2382-1G
A2382-5G
TCA2382-1GEA
66
USD
TCA2382-1G
TCA2382-5G
2-(2-Aminoethyl)-1-methylpyrrolidine ≥97.0% (by GC)
2-(2-Aminoethyl)-1-methylpyrrolidine
CAS Number: 51387-90-7
MDL Number: MFCD00003175
Molecular Formula: C7H16N2
Molecular Weight: 128.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 65
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: 2-(1-Methyl-2-pyrrolidinyl)ethylamine
Formula:
C₇H₁₆N₂ MW: 128.22 g/mol Density: 0.91 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00003175 CAS Number: 51387-90-7 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...