Amantadine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Adamantylamine, 1-Aminotricyclo[3.3.1.1(3,7)]decane, Adamantan-1-amine, 1-Adamantanamine, 1-Aminoadamantane

A2318-5G A2318-25G
TCA2318-5GEA 82.43 USD
TCA2318-5G 76218-742
Amantadine ≥98.0% (by GC, titration analysis)
Amantadine

CAS Number: 768-94-5
MDL Number: MFCD00074732
Molecular Formula: C10H17N
Molecular Weight: 151.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 209


Other Product Names / Synonyms: 1-Aminoadamantane, Amantadine

Formula: C₁₀H₁₇N
MW: 151.25 g/mol
Melting Pt: 209 °C
MDL Number: MFCD00074732
CAS Number: 768-94-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22ba7488f9b0bda3b9fc934db0d230350d0b7ac0&vid=22ba7488f9b0bda3b9fc934db0d230350d0b7ac0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR