5-Aza-2'-deoxycytidine

Supplier: TCI America

Synonyms: 5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one

A2232-100MG A2232-20MG
TCA2232-100MGEA 256 USD
TCA2232-100MG TCA2232-20MG
5-Aza-2'-deoxycytidine
5-Aza-2'-deoxycytidine

CAS Number: 2353-33-5
MDL Number: MFCD00043011
Molecular Formula: C8H12N4O4
Molecular Weight: 228.21
Form: Crystal
Color: White
Melting point (°C): 191
Specific rotation [a]20/D: 68.5 deg (C=0.5, H2O)
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, Decitabine

Formula: C₈H₁₂N₄O₄
Melting Pt: 191 °C
MDL Number: MFCD00043011
CAS Number: 2353-33-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440265e4bbf5316644e8b6b1208c0127c0f426f9&vid=440265e4bbf5316644e8b6b1208c0127c0f426f9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR