AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride) ≥99.0% (by HPLC) for biochemical research

Supplier: TCI America

Synonyms: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride

A2215-100MG A2215-1G
TCA2215-100MGEA 90.19 USD
TCA2215-100MG TCA2215-1G
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride) ≥99.0% (by HPLC) for biochemical research
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulphonylfluoride hydrochloride)

[for Biochemical Research]
CAS Number: 30827-99-7
MDL Number: MFCD00132962
Molecular Formula: C8H10FNO2S
Molecular Weight: 239.69
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 183
Storage Temperature: <0°C


Product Categories: Bioscience, Protease Inhibitors (for Biochemical Research)
Other Product Names / Synonyms: AEBSF

Formula: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00132962
CAS Number: 30827-99-7
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87debf94ee7912e3827bd2367fea6fceaa57df03&vid=a802540dac5349bd626e4d00f0197eea427e8697&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR