3-Amino-9-ethyl carbazole (AEC) ≥90.0% (by HPLC, titration analysis) for biochemical research

Supplier: TCI America

Synonyms: 9-Ethylcarbazol-3-ylamine, 9-Ethyl-9H-carbazol-3-amine, AEC, 50X, AEC, 3-Amino-9-ethylcarbazole

A2167-1G A2167-5G
TCA2167-1GEA 32 USD
TCA2167-1G TCA2167-5G
3-Amino-9-ethyl carbazole (AEC) ≥90.0% (by HPLC, titration analysis) for biochemical research
3-Amino-9-ethyl carbazole (AEC)

[for Biochemical Research]
CAS Number: 132-32-1
MDL Number: MFCD00004964
Molecular Formula: C14H14N2
Molecular Weight: 210.28
Purity/Analysis Method: >90.0% (HPLC,T)
Form: Crystal
Melting point (°C): 103


Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: AEC

Formula: C₁₄H₁₄N₂
MW: 210.28 g/mol
Melting Pt: 103 °C
MDL Number: MFCD00004964
CAS Number: 132-32-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=baba704ce8709af5f358fe01fa6d8c9dd475b1b2&vid=115eabc0cd5bc5bf0adf301f697a2825df7f9d92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR