Alendronic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Amino-1-hydroxybutane-1,1-diphosphonic Acid, 4-Amino-1-hydroxybutane-1,1-diphosphonic acid, Alendronic Acid

A2120-25G A2120-5G
TCA2120-25GEA 215.54 USD
TCA2120-25G TCA2120-5G
Alendronic acid ≥98.0% (by titrimetric analysis)
Alendronic acid

CAS Number: 66376-36-1
MDL Number: MFCD00868112
Molecular Formula: C4H13NO7P2
Molecular Weight: 249.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 235


Product Categories: Bioscience, Anti-osteoporosis Ingredients
Other Product Names / Synonyms: 4-Amino-1-hydroxybutane-1,1-diphosphonic Acid

Formula: C₄H₁₃NO₇P₂
MW: 249.10 g/mol
Melting Pt: 235 °C
MDL Number: MFCD00868112
CAS Number: 66376-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abad256f0a7d09ee3044e35fa5a35e3bd741c95e&vid=abad256f0a7d09ee3044e35fa5a35e3bd741c95e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR