4-Amino-3-methylbenzonitrile ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Cyano-2-methylaniline
A2107-1G
A2107-5G
TCA2107-1GEA
47
USD
TCA2107-1G
TCA2107-5G
4-Amino-3-methylbenzonitrile ≥97.0% (by GC, titration analysis)
4-Amino-3-methylbenzonitrile
CAS Number: 78881-21-7
MDL Number: MFCD02093969
Molecular Formula: C8H8N2
Molecular Weight: 132.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 93
Other Product Names / Synonyms: 4-Cyano-o-toluidine, 4-Cyano-2-methylaniline, 2-Amino-5-cyanotoluene
Formula:
C₈H₈N₂ MW: 132.16 g/mol Melting Pt: 93 °C |
MDL Number:
MFCD02093969 CAS Number: 78881-21-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...