4-Amino-1-methylpiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A2046-25G A2046-5G
TCA2046-25GEA 165.3 USD
TCA2046-25G TCA2046-5G
4-Amino-1-methylpiperidine ≥98.0% (by GC, titration analysis)
4-Amino-1-methylpiperidine

CAS Number: 41838-46-4
MDL Number: MFCD01570549
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 83
Specific Gravity (20/20): 0.91

Formula: C₆H₁₄N₂
MW: 114.1888 g/mol
Boiling Pt: 83 °C
Density: 0.91 (20 °C)
MDL Number: MFCD01570549
CAS Number: 41838-46-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7258e3235aece84eb53ef108a69c0ce8de9dd12&vid=b7258e3235aece84eb53ef108a69c0ce8de9dd12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR