2-(2-Aminoethyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine
A1999-25G
A1999-5G
TCA1999-25GEA
193
USD
TCA1999-25G
TCA1999-5G
2-(2-Aminoethyl)pyridine ≥98.0% (by GC, titration analysis)
2-(2-Aminoethyl)pyridine
CAS Number: 2706-56-1
MDL Number: MFCD00006367
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 100
Specific Gravity (20/20): 1.03
Other Product Names / Synonyms: 2-(2-Pyridyl)ethylamine
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Boiling Pt: 93 °C Density: 1.03 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00006367 CAS Number: 2706-56-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...