(S)-(-)-3-Amino-1,2-propanediol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-3-Amino-1,2-propanediol, (S)-3-Aminopropane-1,2-diol

A1980-1G A1980-5G
TCA1980-1GEA 57.5 USD
TCA1980-1G TCA1980-5G
(S)-(-)-3-Amino-1,2-propanediol ≥98.0% (by GC, titration analysis)
(S)-(-)-3-Amino-1,2-propanediol

CAS Number: 61278-21-5
MDL Number: MFCD00798260
Molecular Formula: C3H9NO2
Molecular Weight: 91.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 55
Flash Point (°C): 113
Specific rotation [a]20/D: -3 deg (C=5, H2O)

Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 130 °C
Melting Pt: 55 °C
Flash Pt: 113 °C
MDL Number: MFCD00798260
CAS Number: 61278-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36d33d3f1233ac51629e19fc1d3791659c300886&vid=36d33d3f1233ac51629e19fc1d3791659c300886&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR