(R)-(+)-3-Amino-1,2-propanediol ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 66211-46-9
MDL Number: MFCD00798261
Molecular Formula: C3H9NO2
Molecular Weight: 91.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 163
Melting point (°C): 55
Flash Point (°C): 113
Specific rotation [a]20/D: 3 deg (C=5, H2O)
Formula:
C₃H₉NO₂ Boiling Pt: 163 °C Melting Pt: 55 °C Flash Pt: 113 °C |
MDL Number:
MFCD00798261 CAS Number: 66211-46-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...