4-Amino-3-fluorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
A1965-25G
A1965-5G
TCA1965-25GEA
622
USD
TCA1965-25G
TCA1965-5G
4-Amino-3-fluorobenzonitrile ≥98.0% (by GC)
4-Amino-3-fluorobenzonitrile
CAS Number: 63069-50-1
MDL Number: MFCD00055559
Molecular Formula: C7H5FN2
Molecular Weight: 136.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 88
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-2-fluoroaniline
Formula:
C₇H₅FN₂ MW: 136.13 g/mol Melting Pt: 88 °C |
MDL Number:
MFCD00055559 CAS Number: 63069-50-1 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...