2-Amino-5-bromobenzonitrile ≥98.0%
Supplier: TCI America
Synonyms:
4-Bromo-2-cyanoaniline
A1960-1G
A1960-5G
TCA1960-1GEA
38
USD
TCA1960-1G
TCA1960-5G
2-Amino-5-bromobenzonitrile ≥98.0%
2-Amino-5-bromobenzonitrile
CAS Number: 39263-32-6
MDL Number: MFCD00158946
Molecular Formula: C7H5BrN2
Molecular Weight: 197.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 96
Other Product Names / Synonyms: 4-Bromo-2-cyanoaniline, 5-Bromoanthranilonitrile
Formula:
C₇H₅BrN₂ Melting Pt: 96 °C |
MDL Number:
MFCD00158946 CAS Number: 39263-32-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...