3-Amino-4-methylpyridine ≥98.0%

Supplier: TCI America

A1957-25G A1957-5G
TCA1957-25GEA 150.6 USD
TCA1957-25G TCA1957-5G
3-Amino-4-methylpyridine ≥98.0%
3-Amino-4-methylpyridine

CAS Number: 3430-27-1
MDL Number: MFCD00128871
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 254
Melting point (°C): 106


Other Product Names / Synonyms: 3-Amino-4-picoline

Formula: C₆H₈N₂
MW: 108.14
Boiling Pt: 254 °C
Melting Pt: 106 °C
MDL Number: MFCD00128871
CAS Number: 3430-27-1
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f530fc81f55a6abb548d1e5b304270141f289642&vid=f530fc81f55a6abb548d1e5b304270141f289642&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR