4-Amino-3,5-dichlorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 78473-00-4
MDL Number: MFCD00017341
Molecular Formula: C7H4Cl2N2
Molecular Weight: 187.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 119
Other Product Names / Synonyms: 2,6-Dichloro-4-cyanoaniline
Formula:
C₇H₄Cl₂N₂ Melting Pt: 119 °C |
MDL Number:
MFCD00017341 CAS Number: 78473-00-4 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...