2-Amino-5-bromopyrimidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Bromopyrimidin-2-ylamine, 5-Bromopyrimidin-2-amine

A1920-25G A1920-5G
TCA1920-25GEA 179.9 USD
TCA1920-25G TCA1920-5G
2-Amino-5-bromopyrimidine ≥98.0% (by GC, titration analysis)
2-Amino-5-bromopyrimidine

CAS Number: 7752-82-1 Molecular Formula: C4H4BrN3 Molecular Weight: 174.00 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 244

Formula: C₄H₄BrN₃
MW: 174 g/mol
Melting Pt: 244 °C
MDL Number: MFCD00012341
CAS Number: 7752-82-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceb288d61fab61e4013c7f47269e22158019e2ff&vid=ceb288d61fab61e4013c7f47269e22158019e2ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR