o-Acetoxybenzoyl chloride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Chlorocarbonyl)phenyl acetate, O-Acetylsalicyloyl chloride, 2-Acetylsalicyloyl chloride, 2-Acetoxybenzoyl chloride

A1917-25G A1917-5G
TCA1917-25GEA 113 USD
TCA1917-25G TCA1917-5G
o-Acetoxybenzoyl chloride ≥97.0% (by HPLC, titration analysis)
o-Acetoxybenzoyl chloride

CAS Number: 5538-51-2
MDL Number: MFCD00000663
Molecular Formula: C9H7ClO3
Molecular Weight: 198.60
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 49
Flash Point (°C): 113


Other Product Names / Synonyms: 2-Acetoxybenzoyl Chloride

Formula: C₉H₇ClO₃
MW: 198.61 g/mol
Boiling Pt: 135 °C
Melting Pt: 49 °C
Flash Pt: 113 °C
MDL Number: MFCD00000663
CAS Number: 5538-51-2
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c62b5601f27e7a1e0135c1550a577f641f0c170d&vid=c62b5601f27e7a1e0135c1550a577f641f0c170d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR