3-Aminothiophenol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Aminophenyl mercaptan, 3-Mercaptoaniline, 3-Aminobenzenethiol

A1852-25G A1852-5G
TCA1852-25GEA 342 USD
TCA1852-25G TCA1852-5G
3-Aminothiophenol ≥97.0% (by GC, titration analysis)
3-Aminothiophenol

CAS Number: 22948-02-3
MDL Number: MFCD00007791
Molecular Formula: C6H7NS
Molecular Weight: 125.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 79
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: 3-Aminothiophenol, 3-Mercaptoaniline

Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 190 °C
Density: 1.19 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00007791
CAS Number: 22948-02-3
UN: 3267
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc29f9fe199def40cd3fb4631e48c4d3b95be889&vid=fc29f9fe199def40cd3fb4631e48c4d3b95be889&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR