6-Aminonicotinonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-Amino-3-pyridinecarbonitrile, 6-aminopyridine-3-carbonitrile, 2-Amino-5-cyanopyridine, 2-Aminopyridine-5-carbonitrile

A1642-25G A1642-5G
TCA1642-25GEA 407 USD
TCA1642-25G TCA1642-5G
6-Aminonicotinonitrile ≥98.0% (by GC, titration analysis)
6-Aminonicotinonitrile

CAS Number: 4214-73-7
MDL Number: MFCD00128902
Molecular Formula: C6H5N3
Molecular Weight: 119.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 250
Melting point (°C): 164


Other Product Names / Synonyms: 6-Aminonicotinonitrile, 6-Amino-3-pyridinecarbonitrile

Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 250 °C
Melting Pt: 164 °C
MDL Number: MFCD00128902
CAS Number: 4214-73-7
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ac33844eb1d38cda9352e637a39957e765c3742&vid=2ac33844eb1d38cda9352e637a39957e765c3742&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR