Thiophene-2-ethylamine ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 30433-91-1
MDL Number: MFCD00051495
Molecular Formula: C6H9NS
Molecular Weight: 127.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 88
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: 2-(2-Thienyl)ethylamine
Formula:
C₆H₉NS Boiling Pt: 201 °C Density: 1.09 (20 °C) Flash Pt: 88 °C |
MDL Number:
MFCD00051495 CAS Number: 30433-91-1 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...