Trimethylsilylmethyl acetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (Trimethylsilyl)methyl acetate, Acetic Acid Trimethylsilylmethyl Ester

A1613-25G A1613-5G
TCA1613-25GEA 113 USD
TCA1613-25G TCA1613-5G
Trimethylsilylmethyl acetate ≥98.0% (by GC)
Trimethylsilylmethyl acetate

CAS Number: 2917-65-9
MDL Number: MFCD00009650
Molecular Formula: C6H14O2Si
Molecular Weight: 146.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 20
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Acetic Acid Trimethylsilylmethyl Ester

Formula: C₆H₁₄O₂Si
MW: 146.26 g/mol
Boiling Pt: 137 °C
Density: 0.88 (20 °C)
Flash Pt: 20 °C
MDL Number: MFCD00009650
CAS Number: 2917-65-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1bc4f314750bb6445384c76f8a1686ba908e019&vid=f1bc4f314750bb6445384c76f8a1686ba908e019&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR