(4-Aminophenyl)acetonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(4-Aminophenyl)acetonitrile, 4-Aminobenzyl cyanide

A1612-25G A1612-5G
TCA1612-25GEA 152 USD
TCA1612-25G TCA1612-5G
(4-Aminophenyl)acetonitrile ≥98.0% (by GC, titration analysis)
(4-Aminophenyl)acetonitrile

CAS Number: 3544-25-0
MDL Number: MFCD00007912
Molecular Formula: C8H8N2
Molecular Weight: 132.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 46
Flash Point (°C): 110


Other Product Names / Synonyms: (4-Aminophenyl)acetonitrile

Formula: C₈H₈N₂
MW: 132.17 g/mol
Melting Pt: 46 °C
Flash Pt: 110 °C
MDL Number: MFCD00007912
CAS Number: 3544-25-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c1686c2aa471d24e8d3291df75a69757e842db5&vid=8c1686c2aa471d24e8d3291df75a69757e842db5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR