Allylpentafluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-(Pentafluorophenyl)-1-propene, 1-Allyl-2,3,4,5,6-pentafluorobenzene

A1611-25G A1611-5G
TCA1611-25GEA 137 USD
TCA1611-25G TCA1611-5G
Allylpentafluorobenzene ≥98.0% (by GC)
Allylpentafluorobenzene

CAS Number: 1736-60-3
MDL Number: MFCD00000301
Molecular Formula: C9H5F5
Molecular Weight: 208.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 42
Specific Gravity (20/20): 1.35


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers, Monomers
Other Product Names / Synonyms: 2,3,4,5,6-Pentafluoro(allylbenzene)

Formula: (C₆F₅)CH₂CHCH₂
MW: 208.13 g/mol
Density: 1.35 (20 °C)
Flash Pt: 42 °C
MDL Number: MFCD00000301
CAS Number: 1736-60-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf4a2231dca12a6a02dfaa9c464b8d0e24891642&vid=4c4252e1b4c19e363facb2be89bd3d570ee4ab0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR