Azobenzene-4,4'-dicarbonyl dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4,4'-Azodibenzoyl Dichloride, Azobenzene-4,4'-dicarbonyl Dichloride

A1575-1G A1575-5G
TCA1575-1GEA 136 USD
TCA1575-1G TCA1575-5G
Azobenzene-4,4'-dicarbonyl dichloride ≥98.0% (by titrimetric analysis)
Azobenzene-4,4'-dicarbonyl dichloride

CAS Number: 10252-29-6
MDL Number: MFCD00053223
Molecular Formula: C14H8Cl2N2O2
Molecular Weight: 307.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 164


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4,4'-Azodibenzoyl Dichloride

Formula: C₁₄H₈Cl₂N₂O₂
MW: 307.13 g/mol
Melting Pt: 164 °C
MDL Number: MFCD00053223
CAS Number: 10252-29-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10bb0fae2d1085ecbba17cc49f74e03c9c4c477d&vid=10bb0fae2d1085ecbba17cc49f74e03c9c4c477d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR