2-Amino-5-chlorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 5922-60-1
MDL Number: MFCD00017106
Molecular Formula: C7H5ClN2
Molecular Weight: 152.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 96
Flash Point (°C): 110
Other Product Names / Synonyms: 5-Chloroanthranilonitrile
Formula:
C₇H₅ClN₂ Boiling Pt: 159 °C Melting Pt: 96 °C Flash Pt: 110 °C |
MDL Number:
MFCD00017106 CAS Number: 5922-60-1 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...