2,1,3-Benzothiadiazol-4-ylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Aminobenzo-2,1,3-thiadiazole, 4-Aminopiazthiole, Benzo[c][1,2,5]thiadiazol-4-amine, 4-Amino-2,1,3-benzothiadiazole
A1385-1G
A1385-5G
TCA1385-1GEA
83
USD
TCA1385-1G
TCA1385-5G
2,1,3-Benzothiadiazol-4-ylamine ≥98.0% (by GC, titration analysis)
2,1,3-Benzothiadiazol-4-ylamine
CAS Number: 767-64-6
MDL Number: MFCD00005810
Molecular Formula: C6H5N3S
Molecular Weight: 151.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 68
Other Product Names / Synonyms: 4-Aminopiazthiole
Formula:
C₆H₅N₃S MW: 151.19 g/mol Melting Pt: 68 °C |
MDL Number:
MFCD00005810 CAS Number: 767-64-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...