N-Boc-1,3-propanediamine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
N-(3-Aminopropyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(3-Aminopropyl)carbamate, N-Boc-1,3-propanediamine, tert-Butyl (3-aminopropyl)carbamate, N-1-Boc-1,3-diaminopropane·HCl, N-(tert-Butoxycarbonyl)-1,3-propanediamine, N-Boc-1,3-diaminopropane
CAS Number: 75178-96-0
MDL Number: MFCD00210021
Molecular Formula: C8H18N2O2
Molecular Weight: 174.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 203
Flash Point (°C): 110
Freezing point (°C): 23
Product Categories: Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: N-(3-Aminopropyl)carbamic Acid tert-Butyl Ester, N-Boc-1,3-diaminopropane, N-Boc-1,3-propanediamine, N-(tert-Butoxycarbonyl)-1,3-propanediamine, tert-Butyl N-(3-Aminopropyl)carbamate
Formula:
C8H18N2O2 MW: 174.244 g/mol Boiling Pt: 203 °C Flash Pt: 110 °C |
MDL Number:
MFCD01076598 CAS Number: 75178-96-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...