N-Boc-ethylenediamine ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
N-(tert-Butoxycarbonyl)-1,2-diaminoethane, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl (2-aminoethyl)carbamate, t-boc-N-EDA, tert-Butyl N-(2-Aminoethyl)carbamate, N-BOC-1,2-diaminoethane
CAS Number: 57260-73-8
MDL Number: MFCD00191871
Molecular Formula: C7H16N2O2
Molecular Weight: 160.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 110
Specific Gravity (20/20): 1.01
Product Categories: Bioscience, Chemical Building Blocks for Polyamines, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: N-(2-Aminoethyl)carbamic Acid tert-Butyl Ester, N-Boc-1,2-diaminoethane, N-Boc-ethylenediamine, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl N-(2-Aminoethyl)carbamate
Formula:
C₇H₁₆N₂O₂ MW: 160.22 g/mol Boiling Pt: 105 °C Density: 1.01 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00191871 CAS Number: 57260-73-8 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...