3-(2-Aminoethyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(Pyridin-3-yl)ethanamine, 2-(3-Pyridyl)ethylamine

A1336-25G A1336-5G
TCA1336-25GEA 514 USD
TCA1336-25G TCA1336-5G
3-(2-Aminoethyl)pyridine ≥98.0% (by GC, titration analysis)
3-(2-Aminoethyl)pyridine

CAS Number: 20173-24-4
MDL Number: MFCD00191604
Molecular Formula: C7H10N2
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 128
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 2-(3-Pyridyl)ethylamine

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 128 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00191604
CAS Number: 20173-24-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=385806887e683cb14acbd76697c92dda58d57473&vid=385806887e683cb14acbd76697c92dda58d57473&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR