2-Amino-1,3-propanediol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol

A1252-25G A1252-5G
TCA1252-25GEA 155.9 USD
TCA1252-25G TCA1252-5G
2-Amino-1,3-propanediol ≥98.0% (by GC, titration analysis)
2-Amino-1,3-propanediol

CAS Number: 534-03-2
MDL Number: MFCD00051487
Molecular Formula: C3H9NO2
Molecular Weight: 91.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 116
Melting point (°C): 52
Flash Point (°C): 110


Other Product Names / Synonyms: Serinol

Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 116 °C
Melting Pt: 52 °C
Flash Pt: 110 °C
MDL Number: MFCD00051487
CAS Number: 534-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbae4806cad4331d67fd8deb1dfdb7423087ac2b&vid=dbae4806cad4331d67fd8deb1dfdb7423087ac2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR