1-Acetoxy-3-chloroacetone ≥95.0% (by GC)

Supplier: TCI America

A1232-25G A1232-5G
TCA1232-25GEA 386.58 USD
TCA1232-25G TCA1232-5G
1-Acetoxy-3-chloroacetone ≥95.0% (by GC)
1-Acetoxy-3-chloroacetone

CAS Number: 40235-68-5
MDL Number: MFCD00191365
Molecular Formula: C5H7ClO3
Molecular Weight: 150.56
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: 1-Acetoxy-3-acetenyl Chloride, 1-Acetoxy-3-chloro-2-propanone

Formula: C₅H₇ClO₃
Density: 1.28 (20 °C)
MDL Number: MFCD00191365
CAS Number: 40235-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73a8ff3e5d04992ec0439b6463168ba5f91e649a&vid=73a8ff3e5d04992ec0439b6463168ba5f91e649a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR