Allylcyclohexane 97,0

Supplier: TCI America

A1201-25ML A1201-5ML
TCA1201-25MLEA 259 USD
TCA1201-25ML TCA1201-5ML
Allylcyclohexane 97,0
Allylcyclohexane

CAS Number: 2114-42-3
MDL Number: MFCD00013769
Molecular Formula: C9H16
Molecular Weight: 124.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 29
Specific Gravity (20/20): 0.81


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3-Cyclohexylpropene

Formula: C₉H₁₆
MW: 124.23 g/mol
Boiling Pt: 151 °C
Density: 0.81 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00013769
CAS Number: 2114-42-3
UN: 3295
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcfb03c0b4f81d54a1d06427dadd965fabb186f3&vid=fcfb03c0b4f81d54a1d06427dadd965fabb186f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR