(S)-(-)-3-Aminopyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A1168-25G
A1168-5G
TCA1168-25GEA
699.9
USD
TCA1168-25G
TCA1168-5G
(S)-(-)-3-Aminopyrrolidine ≥98.0% (by GC, titration analysis)
(S)-(-)-3-Aminopyrrolidine
CAS Number: 128345-57-3
MDL Number: MFCD00143193
Molecular Formula: C4H10N2
Molecular Weight: 86.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 68
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: -10 deg (C=10, H2O)
Formula:
C₄H₁₀N₂ MW: 86.14 g/mol Density: 0.98 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00143193 CAS Number: 128345-57-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...