2-Acetoxyisobutyroyl chloride ≥97.0%

Supplier: TCI America

Synonyms: α-Acetoxyisobutyryl chloride,2-Acetoxy-2-methylpropionyl chloride, 1-Chloro-2-methyl-1-oxopropan-2-yl acetate, 2-Acetoxyisobutyryl Chloride, Chlorocarbonyl-1-methylethyl acetate

A1120-25G A1120-5G
TCA1120-25GEA 144 USD
TCA1120-25G TCA1120-5G
2-Acetoxyisobutyroyl chloride ≥97.0%
2-Acetoxyisobutyroyl chloride

CAS Number: 40635-66-3
MDL Number: MFCD00000708
Molecular Formula: C6H9ClO3
Molecular Weight: 164.59
Purity/Analysis Method: >97.0% (T)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 68
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: 2-Acetoxy-2-methylpropionyl Chloride

Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 70 °C
Density: 1.15 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00000708
CAS Number: 40635-66-3
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bfecc9493f80070973847e3bd49440151a9564c&vid=2bfecc9493f80070973847e3bd49440151a9564c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR