Aminomalononitrile-p-toluenesulfonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Aminomalononitrile p-Toluenesulphonate, Aminomalononitrile p-Toluenesulphonic acid, Aminomalononitrile p-Toluenesulfonic acid, Dicyanomethylammonium p-Toluenesulphonate, 2-Aminomalononitrile 4-methylbenzenesulphonate, Dicyanomethylammonium p-Toluenesulfonate
CAS Number: 5098-14-6
MDL Number: MFCD00012848
Molecular Formula: C3H3N3
Molecular Weight: 253.28
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 164
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Aminomalononitrile p-Toluenesulfonic Acid, Dicyanomethylammonium p-Toluenesulfonate
Formula:
C₃H₃N₃ MW: 253.28 g/mol Melting Pt: 164 °C |
MDL Number:
MFCD00012848 CAS Number: 5098-14-6 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...