Allyl methyl sulfide ≥97.0% (by GC)
Supplier: TCI America
CAS Number: 10152-76-8
MDL Number: MFCD00008657
Molecular Formula: C4H8S
Molecular Weight: 88.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 18
Specific Gravity (20/20): 0.88
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3-Methylthio-1-propene
Formula:
C₄H₈S Boiling Pt: 92 °C Density: 0.88 (20 °C) Flash Pt: 18 °C |
MDL Number:
MFCD00008657 CAS Number: 10152-76-8 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...