1-Ethylpropylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1115-25ML A1115-100ML
TCA1115-25MLEA 98.92 USD
TCA1115-25ML TCA1115-100ML
1-Ethylpropylamine ≥98.0% (by GC, titration analysis)
1-Ethylpropylamine

CAS Number: 616-24-0
MDL Number: MFCD00008096
Molecular Formula: C5H13N
Molecular Weight: 87.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 2
Specific Gravity (20/20): 0.75


Other Product Names / Synonyms: 3-Pentylamine

Formula: C₅H₁₃N
Boiling Pt: 91 °C
Density: 0.75 (20 °C)
Flash Pt: 2 °C
MDL Number: MFCD00008096
CAS Number: 616-24-0
UN: 1106
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=189b3ce54959881c641fd75424614f466dab5d21&vid=189b3ce54959881c641fd75424614f466dab5d21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR