1-Aminocyclopentanecarboxylic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1-Aminocyclopentanecarboxylic Acid, Cycloleucine, 1-Aminocyclopentane-1-carboxylic acid
A1063-25G
A1063-5G
A1063-1G
TCA1063-25GEA
307
USD
TCA1063-25G
TCA1063-5G
TCA1063-1G
1-Aminocyclopentanecarboxylic acid ≥98.0% (by titrimetric analysis)
1-Aminocyclopentanecarboxylic acid
CAS Number: 52-52-8
MDL Number: MFCD00001381
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 330
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: Cycloleucine
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Melting Pt: 330 °C |
MDL Number:
MFCD00001381 CAS Number: 52-52-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...