Methyl dichloromethoxyacetate ≥95.0% (by GC)

Supplier: TCI America

A1046-25G A1046-5G
TCA1046-25GEA 245 USD
TCA1046-25G TCA1046-5G
Methyl dichloromethoxyacetate ≥95.0% (by GC)
Methyl dichloromethoxyacetate

CAS Number: 17640-25-4
MDL Number: MFCD00012202
Molecular Formula: C4H6Cl2O3
Molecular Weight: 172.99
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Flash Point (°C): 91
Specific Gravity (20/20): 1.37


Other Product Names / Synonyms: Dichloromethoxyacetic Acid Methyl Ester

Formula: C₄H₆Cl₂O₃
Boiling Pt: 181 °C
Density: 1.37 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00012202
CAS Number: 17640-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bccf2e2b76d2896e2ce17cc69fee670319474d7&vid=9bccf2e2b76d2896e2ce17cc69fee670319474d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR