Cyclobutylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Cyclobutylamine, Cyclobutanamine, Aminocyclobutane

A1036-1ML A1036-5ML
TCA1036-1MLEA 45.77 USD
TCA1036-1ML TCA1036-5ML
Cyclobutylamine ≥97.0% (by GC, titration analysis)
Cyclobutylamine

CAS Number: 2516-34-9
MDL Number: MFCD00001328
Molecular Formula: C4H9N
Molecular Weight: 71.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): -1
Specific Gravity (20/20): 0.83
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Cyclobutylamine

Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 82 °C
Density: 0.83 (20 °C)
Flash Pt: –1 °C
MDL Number: MFCD00001328
CAS Number: 2516-34-9
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a7ff2475974e8239d03c74d92ef2e854aad6bcb&vid=4a7ff2475974e8239d03c74d92ef2e854aad6bcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR