Allyl ethyl ether ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Allyl Ethyl Ether

A1033-25ML A1033-5ML
TCA1033-25MLEA 174 USD
TCA1033-25ML TCA1033-5ML
Allyl ethyl ether ≥95.0% (by GC)
Allyl ethyl ether

CAS Number: 557-31-3
MDL Number: MFCD00009255
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): -20
Specific Gravity (20/20): 0.77

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 67 °C
Density: 0.77 (20 °C)
Flash Pt: -20 °C
MDL Number: MFCD00009255
CAS Number: 557-31-3
UN: 2335
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f994ba93bb6c24ead634c53eaca1720f073f3428&vid=f994ba93bb6c24ead634c53eaca1720f073f3428&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR