1-(2-Aminoethyl)piperidine ≥97.0%
Supplier: TCI America
Synonyms:
N-(2-Aminoethyl)piperidine
A1026-25ML
A1026-5ML
TCA1026-25MLEA
122
USD
TCA1026-25ML
TCA1026-5ML
1-(2-Aminoethyl)piperidine ≥97.0%
1-(2-Aminoethyl)piperidine
CAS Number: 27578-60-5
MDL Number: MFCD00006516
Molecular Formula: C7H16N2
Molecular Weight: 128.22
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 184
Flash Point (°C): 58
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: 2-(1-Piperidino)ethylamine
Formula:
C₇H₁₆N₂ Boiling Pt: 184 °C Density: 0.91 (20 °C) Flash Pt: 58 °C |
MDL Number:
MFCD00006516 CAS Number: 27578-60-5 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...