4-Aminobutan-1-ol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Hydroxybutylamine, 4-Amino-1-butanol

A1013-25G A1013-5G
TCA1013-25GEA 264.51 USD
TCA1013-25G TCA1013-5G
4-Aminobutan-1-ol ≥98.0% (by GC)
4-Aminobutan-1-ol

CAS Number: 13325-10-5
MDL Number: MFCD00008230
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Flash Point (°C): 108
Specific Gravity (20/20): 0.96

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 206 °C
Density: 0.96 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00008230
CAS Number: 13325-10-5
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8210b631a54dec476b9bf4e1d7b8c551f3e36ad1&vid=8210b631a54dec476b9bf4e1d7b8c551f3e36ad1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR