6-Aminonicotinamide ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
6-Aminopyridine-3-carboxamide
A0986-1G
A0986-5G
TCA0986-1GEA
111.93
USD
TCA0986-1G
TCA0986-5G
6-Aminonicotinamide ≥99.0% (by titrimetric analysis)
6-Aminonicotinamide
CAS Number: 329-89-5
MDL Number: MFCD00006327
Molecular Formula: C6H7N3O
Molecular Weight: 137.14
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2-Aminopyridine-5-carboxamide
Formula:
C₆H₇N₃O MW: 137.14 g/mol |
MDL Number:
MFCD00006327 CAS Number: 329-89-5 |
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...