(S)-(+)-1-Aminopropan-2-ol ≥98.0% (by GC)

Supplier: TCI America

A0975-1G A0975-5G
TCA0975-1GEA 40 USD
TCA0975-1G TCA0975-5G
(S)-(+)-1-Aminopropan-2-ol ≥98.0% (by GC)
(S)-(+)-1-Aminopropan-2-ol

CAS Number: 2799-17-9
MDL Number: MFCD00064429
Molecular Formula: C3H9NO
Molecular Weight: 75.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 160
Flash Point (°C): 78
Freezing point (°C): 22
Specific rotation [a]20/D: 24 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-Isopropanolamine

Formula: C₃H₉NO
Boiling Pt: 160 °C
Flash Pt: 78 °C
MDL Number: MFCD00064429
CAS Number: 2799-17-9
UN: 3259
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5466751f4908f7cb816bd83e631a55a420015625&vid=5466751f4908f7cb816bd83e631a55a420015625&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR